Assay Details
| Gene Symbol: LOC102467213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104671.1 | 3 - 4 | 219 | 95 | ||
| GenBank mRNA | AA292843.1 | - | 3 - 4 | 219 | 95 | |
| AA398661.1 | - | 1 - 2 | 95 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB9B, member RAS oncogene family pseudogene 1 |
RAB9BP1 | RAB9P1 | -1.7mb | Anti-sense | Chr. 5 - 105099474 - 105100098 on NCBI Build GRCh38 |
| Downstream | ephrin A5 | EFNA5 |
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS |
558.4kb | Sense | Chr. 5 - 107376889 - 107672364 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102467213
Genomic Map