Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016630.hg.1 |
TC08002584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016630.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016630.hg.1 |
ENST00000421742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016630.hg.1 |
ENST00000522909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016630.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016634.hg.1 |
TC08002584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016634.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016634.hg.1 |
ENST00000421742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016634.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016629.hg.1 |
TC08002584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016629.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016629.hg.1 |
ENST00000421742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016629.hg.1 |
ENST00000522909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016629.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187241.hg.1 |
TC0800012423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000520633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000520633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187247.hg.1 |
TC0800012423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187242.hg.1 |
TC0800012423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000324682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000521581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000522909.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
ENST00000523545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
NM_001013626.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187243.hg.1 |
TC0800012423.hg.1 |
Clariom D TC |