Assay Details
| Gene Symbol: MYCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293233.1 | NP_001280162.1 | 1 - 2 | 302 | 124 | |
| NM_005378.5 | NP_005369.2 | 1 - 2 | 302 | 124 | ||
| GenBank mRNA | AF320053.1 | - | 1 - 2 | 176 | 124 | |
| AK298225.1 | - | 1 - 2 | 182 | 124 | ||
| AK299495.1 | - | 1 - 2 | 176 | 124 | ||
| BU167361.1 | - | 210 | 124 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000094.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK298225.1,AK299495.1,NM_005378.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cartilage condensation
positive regulation of mesenchymal cell proliferation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of cell death
lung development
embryonic digit morphogenesis
regulation of auditory receptor cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
negative regulation of astrocyte differentiation
branching morphogenesis of an epithelial tube
negative regulation of reactive oxygen species metabolic process
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein dimerization activity
cartilage condensation
positive regulation of mesenchymal cell proliferation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of cell death
lung development
embryonic digit morphogenesis
regulation of auditory receptor cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
negative regulation of astrocyte differentiation
branching morphogenesis of an epithelial tube
negative regulation of reactive oxygen species metabolic process
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001175.hg.1 | TC02000094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001175.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001175.hg.1 | ENST00000426211.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001175.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001173.hg.1 | TC02000094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001173.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001173.hg.1 | ENST00000426211.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001173.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001176.hg.1 | TC02000094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001176.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001176.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146612.hg.1 | TC0200006799.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006799.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | ENST00000281043.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_001293228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_001293233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | NM_005378.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146613.hg.1 | TC0200006799.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000419083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000420452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000439180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | ENST00000448719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_026766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_110230.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016634.hg.1 | NR_110230.1 | Refseq |

MYCN
Genomic Map