Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013300.2 | NP_037432.2 | 2 - 3 | 630 | 154 | |
| XM_005253875.4 | XP_005253932.1 | 2 - 3 | 140 | 154 | ||
| XM_011538246.2 | XP_011536548.1 | 2 - 3 | 158 | 154 | ||
| GenBank mRNA | AK225731.1 | - | 2 - 3 | 621 | 154 | |
| AK297684.1 | - | 2 - 3 | 140 | 154 | ||
| AK308802.1 | - | 2 - 3 | 243 | 154 | ||
| BC020967.1 | - | 2 - 3 | 446 | 3848037 | 154 | |
| CR457001.1 | - | 2 - 3 | 181 | 154 | ||
| U79274.1 | - | 2 - 3 | 597 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000857.hg
Additional Information:
This assay is in the 5'UTR region of the AK297684.1,XM_005253875.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011179.hg.1 | ENST00000377673.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011179.hg.1 | ENST00000538285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011179.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011179.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011179.hg.1 | TC12000857.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000377673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000538285.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | ENST00000548869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | NM_013300.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165540.hg.1 | TC1200008843.hg.1 | Clariom D TC |

FAM216A
Genomic Map