Assay Details
| Gene Symbol: ATP9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525965.2 | XP_011524267.1 | 2 - 3 | 420 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525965.2 transcripts.
Set Membership: |
|

ATP9B
Genomic Map