| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009015.hg.1 |
ENST00000258168.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009015.hg.1 |
ENST00000425577.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009015.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009015.hg.1 |
TC16000637.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
ENST00000564552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008577.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
ENST00000258168.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
ENST00000563804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
NM_017429.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164002.hg.1 |
TC1600008577.hg.1 |
Clariom D TC |