Assay Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004419.3 | NP_004410.3 | 2 - 3 | 786 | 76 | |
| GenBank mRNA | AB209823.1 | - | 2 - 3 | 732 | 76 | |
| BC062545.1 | - | 2 - 3 | 703 | 5808876 | 76 | |
| U15932.2 | - | 2 - 3 | 738 | 76 | ||
| U16996.1 | - | 2 - 3 | 577 | 76 | ||
More Information
Set Membership: |
|

DUSP5
Genomic Map