Assay Details
| Gene Symbol: HUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004507.3 | NP_004498.1 | 5 - 6 | 637 | 163 | |
| NR_037917.1 | 5 - 6 | 695 | 163 | |||
| XM_017012115.1 | XP_016867604.1 | 5 - 6 | 635 | 163 | ||
| GenBank mRNA | AF076844.1 | - | 5 - 6 | 594 | 163 | |
| AF110393.1 | - | 5 - 6 | 591 | 163 | ||
| AJ227901.1 | - | 1 - 2 | 51 | 164 | ||
| AK290459.1 | - | 5 - 6 | 574 | 163 | ||
| AK294117.1 | - | 5 - 6 | 623 | 163 | ||
| BC007013.1 | - | 5 - 6 | 555 | 3684398 | 163 | |
| BT019481.1 | - | 5 - 6 | 531 | 163 | ||
| BT019482.1 | - | 5 - 6 | 531 | 163 | ||
| CR536552.1 | - | 5 - 6 | 531 | 163 | ||
| Y16893.1 | - | 5 - 6 | 531 | 163 | ||
More Information
Set Membership: |
|

HUS1
Genomic Map