Assay Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016172.2 | NP_057256.2 | 5 - 6 | 764 | 84 | |
| GenBank mRNA | AF038200.1 | - | 4 - 5 | 329 | 84 | |
| AF068195.1 | - | 4 - 5 | 391 | 84 | ||
| AF176796.1 | - | 5 - 6 | 737 | 84 | ||
| BC004967.1 | - | 5 - 6 | 764 | 3535682 | 84 | |
| BC011822.1 | - | 5 - 6 | 590 | 3688526 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001721.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021988.hg.1 | TC09001721.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021988.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021988.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181771.hg.1 | TC0900011959.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |

UBAC1
Genomic Map