Assay Details
| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027129.1 | 2 - 3 | 464 | 92 | ||
| GenBank mRNA | AF380178.1 | - | 2 - 3 | 396 | 92 | |
| AY040088.1 | - | 2 - 3 | 396 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ITGB2 antisense RNA 1 | ITGB2-AS1 | -5.9kb | Anti-sense | Chr. 21 - 44921035 - 44929680 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 207 member A | FAM207A |
C21orf70, PRED56 |
2.3kb | Anti-sense | Chr. 21 - 44939697 - 44976989 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000539.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006510.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006510.hg.1 | ENST00000380070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006510.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006510.hg.1 | TC21000539.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | ENST00000380070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | ENST00000397841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006509.hg.1 | TC21000539.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | ENST00000380070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | ENST00000397841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006511.hg.1 | TC21000539.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006512.hg.1 | ENST00000380070.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006512.hg.1 | ENST00000397841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006512.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006512.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006512.hg.1 | TC21000539.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100156980.hg.1 | TC2100008395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156980.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156980.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156980.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156980.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | TC2100008395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156979.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | ENST00000615847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008395.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | TC2100008395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | ENST00000330551.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156981.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | TC2100008395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | NR_027129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | ENST00000397841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | NR_027128.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156982.hg.1 | NR_027129.1 | Refseq |

LINC01547
Genomic Map