Assay Details
| Gene Symbol: SLC25A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031481.2 | NP_113669.1 | 1 - 2 | 278 | 86 | |
| XM_011546153.2 | XP_011544455.1 | 1 - 2 | 1220 | 86 | ||
| XM_017028969.1 | XP_016884458.1 | 1 - 2 | 1214 | 86 | ||
| XM_017028970.1 | XP_016884459.1 | 1 - 2 | 1214 | 86 | ||
| XM_017028972.1 | XP_016884461.1 | 1 - 2 | 1214 | 86 | ||
| XM_017028974.1 | XP_016884463.1 | 1 - 2 | 1214 | 86 | ||
| XM_017028975.1 | XP_016884464.1 | 1 - 2 | 1214 | 86 | ||
| GenBank mRNA | AJ428203.1 | - | 1 - 2 | 234 | 86 | |
| AK308974.1 | - | 1 - 2 | 232 | 86 | ||
| AY008285.1 | - | 1 - 2 | 234 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000024.hg
Additional Information:
This assay is in the 5'UTR region of the AJ428203.1,AY008285.1,NM_031481.2,XM_011546153.2,XM_017028969.1,XM_017028970.1,XM_017028972.1,XM_017028974.1,XM_017028975.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000150.hg.1 | ENST00000327451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000150.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000150.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000150.hg.1 | TC22000024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000145.hg.1 | ENST00000327451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000145.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000145.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000145.hg.1 | TC22000024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000146.hg.1 | ENST00000327451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000146.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000146.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000146.hg.1 | TC22000024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000153.hg.1 | ENST00000327451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000153.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000153.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000153.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000153.hg.1 | TC22000024.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | TC2200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144550.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | TC2200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144554.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000467228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000469889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | ENST00000497401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006520.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | TC2200006520.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | NM_031481.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000327451.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000399813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | ENST00000496051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | NM_001303484.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144549.hg.1 | NM_031481.2 | Refseq |

SLC25A18
Genomic Map