Assay Details
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC018067.1 | - | 3 - 4 | 442 | 105 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000510528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000510528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000288723.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000330852.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000338176.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000504644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000508274.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000510528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000510528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000511649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | ENST00000630951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001017979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_001159601.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010077.hg.1 | NM_004249.3 | Refseq |

RAB28
Genomic Map