Assay Details
| Gene Symbol: ADAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015840.3 | NP_056655.2 | 7 - 8 | 2659 | 78 | |
| NM_015841.3 | NP_056656.2 | 7 - 8 | 2602 | 78 | ||
| XM_011509061.2 | XP_011507363.2 | 7 - 8 | 2528 | 78 | ||
| XM_017000037.1 | XP_016855526.1 | 8 - 9 | 2456 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ADAR
Genomic Map