| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
ENST00000359191.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
ENST00000378524.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
ENST00000402194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
ENST00000432763.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
NM_001111020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
NM_001130824.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
NM_001130825.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
NM_003169.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007456.hg.1 |
TC19000547.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000359191.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000402194.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000432763.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000593727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598725.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000599117.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000599907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001111020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001130824.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001130825.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_003169.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
TC1900008059.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000359191.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000402194.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000432763.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000593727.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598725.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000598786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000599117.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
ENST00000599907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001111020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001130824.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_001130825.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160714.hg.1 |
NM_003169.3 |
Refseq |