Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142386.2 | NP_001135858.1 | 5 - 6 | 831 | 75 | |
| NM_005391.4 | NP_005382.1 | 5 - 6 | 831 | 75 | ||
| GenBank mRNA | AK301965.1 | - | 5 - 6 | 834 | 75 | |
| BC015948.1 | - | 5 - 6 | 778 | 3922929 | 75 | |
| L42452.1 | - | 5 - 6 | 624 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000118.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
glucose metabolic process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000568479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001586.hg.1 | TC0X000118.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001586.hg.1 | ENST00000379162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001586.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001586.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001586.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000568479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | ENST00000379162.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | ENST00000441463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | NM_001142386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | NM_005391.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146682.hg.1 | TC0X00006816.hg.1 | Clariom D TC |

PDK3
Genomic Map