Assay Details
| Gene Symbol: MAP4K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042600.2 | NP_001036065.1 | 6 - 7 | 556 | 78 | |
| NM_007181.5 | NP_009112.1 | 6 - 7 | 556 | 78 | ||
| XM_011526403.1 | XP_011524705.1 | 6 - 7 | 454 | 78 | ||
| XM_011526404.1 | XP_011524706.1 | 6 - 7 | 454 | 78 | ||
| XM_017026231.1 | XP_016881720.1 | 6 - 7 | 462 | 78 | ||
| GenBank mRNA | BX345201.2 | - | 6 - 7 | 524 | 78 | |
| U66464.1 | - | 6 - 7 | 470 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MAP4K1
Genomic Map