Assay Details
| Gene Symbol: IRAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025243.1 | NP_001020414.1 | 10 - 11 | 1385 | 77 | |
| GenBank mRNA | BC014963.1 | - | 10 - 11 | 1370 | 4906853 | 77 |
| DQ054788.1 | - | 10 - 11 | 1306 | 77 | ||
More Information
Set Membership: |
|

IRAK1
Genomic Map