Assay Details
| Gene Symbol: SYN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006950.3 | NP_008881.2 | 12 - 13 | 2113 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000997.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
neurotransmitter secretion
regulation of neurotransmitter secretion
regulation of synaptic vesicle exocytosis
actin binding
catalytic activity
transporter activity
protein binding
ATP binding
protein kinase binding
calcium-dependent protein binding
chemical synaptic transmission
neurotransmitter secretion
regulation of neurotransmitter secretion
regulation of synaptic vesicle exocytosis
actin binding
catalytic activity
transporter activity
protein binding
ATP binding
protein kinase binding
calcium-dependent protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013222.hg.1 | TC0X000997.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013222.hg.1 | ENST00000295987.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013222.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165236.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165236.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165236.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165236.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165236.hg.1 | TC0X00009576.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000295987.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | ENST00000340666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_006950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009576.hg.1 | NM_133499.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 220615 | - | - |

SYN1
Genomic Map