Assay Details
| Gene Symbol: MICU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152726.2 | NP_689939.1 | 4 - 5 | 532 | 97 | |
| XM_017020433.1 | XP_016875922.1 | 4 - 5 | 335 | 97 | ||
| GenBank mRNA | AK091907.1 | - | 4 - 5 | 294 | 97 | |
| BC031089.1 | - | 4 - 5 | 486 | 5300471 | 97 | |
| BC034965.1 | - | 4 - 5 | 494 | 4824307 | 97 | |
| KU178890.1 | - | 4 - 5 | 464 | 97 | ||
| KU178891.1 | - | 6 - 7 | 660 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000474.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017020433.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial calcium ion transport
calcium ion transmembrane import into mitochondrion
mitochondrial calcium ion homeostasis
positive regulation of mitochondrial calcium ion concentration
negative regulation of mitochondrial calcium ion concentration
calcium ion binding
protein binding
protein heterodimerization activity
mitochondrial calcium ion transport
calcium ion transmembrane import into mitochondrion
mitochondrial calcium ion homeostasis
positive regulation of mitochondrial calcium ion concentration
negative regulation of mitochondrial calcium ion concentration
calcium ion binding
protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13005407.hg.1 | ENST00000382374.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005407.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005407.hg.1 | TC13000474.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156030.hg.1 | TC1300008292.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000382374.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000460488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000468222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000469058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000476895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000478700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000479790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | ENST00000480341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008292.hg.1 | NM_152726.2 | Refseq |

MICU2
Genomic Map