Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018455.5 | NP_060925.2 | 6 - 7 | 1318 | 65 | |
| XM_017023456.1 | XP_016878945.1 | 6 - 7 | 718 | 65 | ||
| GenBank mRNA | AF217515.1 | - | 6 - 7 | 868 | 65 | |
| BC007334.2 | - | 6 - 7 | 1264 | 3830110 | 65 | |
| BQ013821.1 | - | 1 - 2 | 65 | |||
| HY363272.1 | - | 1 - 2 | 65 | |||
| HY374758.1 | - | 1 - 2 | 65 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000299572.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000305850.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000393335.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000428963.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000439957.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000562943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000564669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000566639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000568445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000568445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000568445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000568445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000568445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000569461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000569461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | ENST00000569461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100624.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001100625.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_001270474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008567.hg.1 | NM_018455.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000561808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000562315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011571.hg.1 | ENST00000566390.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 100455 | - | - |

CENPN
Genomic Map