Assay Details
| Gene Symbol: RGS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006480.4 | NP_006471.2 | 2 - 3 | 246 | 101 | |
| XM_005265794.4 | XP_005265851.1 | 2 - 3 | 413 | 101 | ||
| XM_005265795.4 | XP_005265852.1 | 2 - 3 | 418 | 101 | ||
| GenBank mRNA | AK123382.1 | - | 2 - 3 | 247 | 101 | |
| BC014094.2 | - | 2 - 3 | 216 | 4547415 | 101 | |
| BI752372.1 | - | 2 - 3 | 252 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001013.hg.1
Set Membership: |
|

RGS14
Genomic Map