Assay Details
| Gene Symbol: ARR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004312.2 | NP_004303.2 | 13 - 14 | 1059 | 76 | |
| XM_017029518.1 | XP_016885007.1 | 11 - 12 | 1036 | 76 | ||
| GenBank mRNA | AF033105.1 | - | 13 - 14 | 1042 | 76 | |
| BC012096.1 | - | 13 - 14 | 1089 | 4645867 | 76 | |
| CR456849.1 | - | 11 - 12 | 991 | 76 | ||
| EU883571.1 | - | 11 - 12 | 991 | 76 | ||
| U03626.1 | - | 11 - 12 | 991 | 76 | ||
More Information
Set Membership: |
|

ARR3
Genomic Map