Assay Details
| Gene Symbol: ARR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004312.2 | NP_004303.2 | 14 - 15 | 1079 | 72 | |
| XM_017029518.1 | XP_016885007.1 | 12 - 13 | 1056 | 72 | ||
| GenBank mRNA | AF033105.1 | - | 14 - 15 | 1062 | 72 | |
| BC012096.1 | - | 14 - 15 | 1109 | 4645867 | 72 | |
| CR456849.1 | - | 12 - 13 | 1011 | 72 | ||
| EU883571.1 | - | 12 - 13 | 1011 | 72 | ||
| U03626.1 | - | 12 - 13 | 1011 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000374.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004825.hg.1 | TC0X000374.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004825.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004825.hg.1 | ENST00000374480.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004825.hg.1 | ENST00000374495.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004825.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151606.hg.1 | TC0X00007535.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000307959.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000374495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000477379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000480877.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | ENST00000620997.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007535.hg.1 | NM_004312.2 | Refseq |

ARR3
Genomic Map