Assay Details
| Gene Symbol: RHOT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011524975.2 | XP_011523277.1 | 2 - 3 | 533 | 78 | |
| XM_017024822.1 | XP_016880311.1 | 1 - 2 | 305 | 78 | ||
| XM_017024824.1 | XP_016880313.1 | 2 - 3 | 411 | 78 | ||
| XM_017024825.1 | XP_016880314.1 | 2 - 3 | 503 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011524975.2, XM_017024822.1, XM_017024824.1, XM_017024825.1 transcripts.
Set Membership: |
|

RHOT1
Genomic Map