Assay Details
| Gene Symbol: GNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006496.3 | NP_006487.1 | 7 - 8 | 1027 | 167 | |
| GenBank mRNA | AF493907.1 | - | 7 - 8 | 870 | 167 | |
| AK311691.1 | - | 8 - 9 | 890 | 167 | ||
| AK312252.1 | - | 7 - 8 | 956 | 167 | ||
| BC025285.1 | - | 7 - 8 | 980 | 5433334 | 167 | |
| BT019973.1 | - | 7 - 8 | 870 | 167 | ||
| BT019974.1 | - | 7 - 8 | 870 | 167 | ||
| J03005.1 | - | 7 - 8 | 878 | 167 | ||
| J03198.1 | - | 7 - 8 | 932 | 167 | ||
| J03238.1 | - | 7 - 8 | 960 | 167 | ||
| M27543.1 | - | 7 - 8 | 912 | 167 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000947.hg.1
Set Membership: |
|

GNAI3
Genomic Map