Assay Details
| Gene Symbol: NCAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004386.2 | NP_004377.2 | 2 - 3 | 173 | 74 | |
| GenBank mRNA | AB210004.1 | - | 2 - 3 | 117 | 74 | |
| AF026547.1 | - | 1 - 2 | 75 | 74 | ||
| CN282070.1 | - | 2 - 3 | 173 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000370.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
cell adhesion
central nervous system development
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
extracellular matrix structural constituent
calcium ion binding
hyaluronic acid binding
carbohydrate binding
skeletal system development
cell adhesion
central nervous system development
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
extracellular matrix structural constituent
calcium ion binding
hyaluronic acid binding
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19004945.hg.1 | ENST00000252575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004945.hg.1 | ENST00000539499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004945.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004945.hg.1 | TC19000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004946.hg.1 | ENST00000252575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004946.hg.1 | ENST00000539499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004946.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004946.hg.1 | TC19000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004944.hg.1 | ENST00000252575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004944.hg.1 | ENST00000539499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004944.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004944.hg.1 | TC19000370.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000585410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000588231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | ENST00000590187.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007426.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154728.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154728.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154728.hg.1 | TC1900007426.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900154728.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154728.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154726.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154726.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154726.hg.1 | TC1900007426.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900154726.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154726.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154729.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154729.hg.1 | NM_004386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154729.hg.1 | TC1900007426.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900154729.hg.1 | ENST00000252575.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154729.hg.1 | NM_004386.2 | Refseq |

NCAN
Genomic Map