Assay Details
| Gene Symbol: IREB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320941.1 | NP_001307870.1 | 13 - 14 | 1883 | 247 | |
| NM_001320942.1 | NP_001307871.1 | 14 - 15 | 2207 | 247 | ||
| NM_004136.3 | NP_004127.1 | 14 - 15 | 1953 | 247 | ||
| GenBank mRNA | AB209403.1 | - | 14 - 15 | 1887 | 247 | |
| AK027033.1 | - | 13 - 14 | 1717 | 247 | ||
| AK027034.1 | - | 5 - 6 | 571 | 247 | ||
| AK292992.1 | - | 14 - 15 | 1837 | 247 | ||
| BC117481.1 | - | 14 - 15 | 1820 | 40126032 | 247 | |
| BC117483.1 | - | 14 - 15 | 1820 | 40126034 | 247 | |
| BC143948.1 | - | 13 - 14 | 1750 | 247 | ||
| M58511.1 | - | 14 - 15 | 1791 | 247 | ||
| U20180.1 | - | 13 - 14 | 1758 | 247 | ||
More Information
Set Membership: |
|

IREB2
Genomic Map