Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184963.1 | NP_001171892.1 | 7 - 8 | 66 | ||
| NM_173160.2 | NP_775183.1 | 7 - 8 | 66 | |||
| GenBank mRNA | AA759244.1 | - | 2 - 3 | 233 | 66 | |
| AI829935.1 | - | 2 - 3 | 240 | 66 | ||
| BC054876.1 | - | 7 - 8 | 4610529 | 66 | ||
| BG432244.1 | - | 7 - 8 | 66 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000272.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10003123.hg.1 | ENST00000374451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003123.hg.1 | ENST00000458363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003123.hg.1 | ENST00000476166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003123.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003123.hg.1 | TC10000272.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003124.hg.1 | ENST00000374451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003124.hg.1 | ENST00000458363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003124.hg.1 | ENST00000476166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003124.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003124.hg.1 | TC10000272.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150407.hg.1 | TC1000007415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150409.hg.1 | TC1000007415.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 224429 | - | - |

FXYD4
Genomic Map