| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
TC07003392.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
ENST00000265299.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
ENST00000409881.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
ENST00000434909.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
ENST00000458257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
ENST00000509504.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
NM_032222.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003108.hg.1 |
NR_037598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000265299.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000409881.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000458257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
NM_032222.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
NR_037598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000265299.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000409881.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
ENST00000458257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
NM_032222.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
NR_037598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196566.hg.1 |
TC0700013346.hg.1 |
Clariom D TC |