Assay Details
| Gene Symbol: C22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005261781.1 | XP_005261838.1 | 6 | 877 | 71 | |
| XM_005261782.3 | XP_005261839.1 | 6 | 757 | 71 | ||
| XM_005261783.2 | XP_005261840.1 | 5 | 814 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MICAL like 1 | MICALL1 |
MICAL-L1, MIRAB13 |
-1.1kb | Anti-sense | Chr. 22 - 37906148 - 37942458 on NCBI Build GRCh38 |
| Downstream | RNA polymerase II subunit F | POLR2F |
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15 |
9.1kb | Anti-sense | Chr. 22 - 37953663 - 38041915 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

C22orf23
Genomic Map