Assay Details
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102371.1 | NP_001095841.1 | 1 - 2 | 94 | 150 | |
GenBank mRNA | BC040351.1 | - | 1 - 2 | 56 | 3873448 | 150 |
DA409026.1 | - | 1 - 2 | 94 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000710.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC040351.1,NM_001102371.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | ENST00000216187.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | ENST00000397224.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012504.hg.1 | TC22000710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22012513.hg.1 | ENST00000397224.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012513.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012513.hg.1 | TC22000710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22012515.hg.1 | ENST00000397224.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012515.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012515.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012515.hg.1 | TC22000710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | ENST00000216187.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | ENST00000397224.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012505.hg.1 | TC22000710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22012514.hg.1 | ENST00000397224.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22012514.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22012514.hg.1 | TC22000710.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | TC2200008610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162033.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | TC2200008610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162034.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000216187.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000366463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397223.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | ENST00000423980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008610.hg.1 | NM_024955.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162043.hg.1 | TC2200008610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200162043.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162043.hg.1 | NM_001102371.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200162043.hg.1 | ENST00000397224.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200162043.hg.1 | NM_001102371.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 172800 | - | - |
