Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206542.1 | NP_001193471.1 | 5 - 6 | 1101 | 101 | |
| NM_001230.4 | NP_001221.2 | 5 - 6 | 1101 | 101 | ||
| GenBank mRNA | AJ487679.1 | - | 5 - 6 | 804 | 101 | |
| KU177960.1 | - | 4 - 5 | 683 | 101 | ||
| U60519.1 | - | 5 - 6 | 830 | 101 | ||
More Information
Set Membership: |
|

CASP10
Genomic Map