Assay Details
| Gene Symbol: TRIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267301.3 | XP_005267358.1 | 8 - 9 | 2974 | 58 | |
| XM_006720018.3 | XP_006720081.1 | 8 - 9 | 2974 | 58 | ||
| XM_011536389.2 | XP_011534691.1 | 8 - 9 | 2977 | 58 | ||
| XM_017020943.1 | XP_016876432.1 | 8 - 9 | 2962 | 58 | ||
| GenBank mRNA | AK055388.1 | - | 7 - 8 | 1982 | 58 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic vesicle exocytosis
protein ubiquitination
negative regulation of SNARE complex assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of calcium ion-dependent exocytosis
SNARE binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
protein homodimerization activity
synaptic vesicle exocytosis
protein ubiquitination
negative regulation of SNARE complex assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of calcium ion-dependent exocytosis
SNARE binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |

TRIM9
Genomic Map