Assay Details
| Gene Symbol: TRIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536389.2 | XP_011534691.1 | 6 - 7 | 2696 | 88 | |
| XM_017020943.1 | XP_016876432.1 | 6 - 7 | 2681 | 88 | ||
| XM_017020946.1 | XP_016876435.1 | 6 - 7 | 2681 | 88 | ||
| XM_017020948.1 | XP_016876437.1 | 6 - 7 | 2681 | 88 | ||
| GenBank mRNA | AK055388.1 | - | 5 - 6 | 1701 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

TRIM9
Genomic Map