Assay Details
| Gene Symbol: DAGLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006133.2 | NP_006124.1 | 11 - 12 | 1333 | 83 | |
| XM_017018238.1 | XP_016873727.1 | 12 - 13 | 1660 | 83 | ||
| XM_017018239.1 | XP_016873728.1 | 12 - 13 | 1565 | 83 | ||
| XM_017018240.1 | XP_016873729.1 | 12 - 13 | 1660 | 83 | ||
| GenBank mRNA | AB014559.2 | - | 11 - 12 | 1328 | 83 | |
| AY275377.1 | - | 10 - 11 | 1217 | 83 | ||
| BC150176.1 | - | 11 - 12 | 1333 | 40148003 | 83 | |
| BC150195.1 | - | 11 - 12 | 1333 | 40148734 | 83 | |
| BC152453.1 | - | 11 - 12 | 1328 | 8862586 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000538.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006501.hg.1 | ENST00000257215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006501.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006501.hg.1 | TC11000538.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155574.hg.1 | TC1100007818.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |

DAGLA
Genomic Map