Assay Details
| Gene Symbol: DAGLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006133.2 | NP_006124.1 | 14 - 15 | 1630 | 81 | |
| XM_017018238.1 | XP_016873727.1 | 15 - 16 | 1957 | 81 | ||
| XM_017018239.1 | XP_016873728.1 | 15 - 16 | 1862 | 81 | ||
| XM_017018240.1 | XP_016873729.1 | 15 - 16 | 1957 | 81 | ||
| GenBank mRNA | AB014559.2 | - | 14 - 15 | 81 | ||
| AY275377.1 | - | 13 - 14 | 81 | |||
| BC150176.1 | - | 14 - 15 | 40148003 | 81 | ||
| BC150195.1 | - | 14 - 15 | 40148734 | 81 | ||
| BC152453.1 | - | 14 - 15 | 8862586 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000538.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006504.hg.1 | ENST00000257215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006504.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006504.hg.1 | TC11000538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100155577.hg.1 | TC1100007818.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000257215.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | ENST00000540717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007818.hg.1 | NM_006133.2 | Refseq |

DAGLA
Genomic Map