Assay Details
| Gene Symbol: NMNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320511.1 | NP_001307440.1 | 3 - 4 | 490 | 58 | |
| XM_017006329.1 | XP_016861818.1 | 3 - 4 | 470 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320511.1, XM_017006329.1 transcripts.
Set Membership: |
|

NMNAT3
Genomic Map