Assay Details
| Gene Symbol: PRRC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025675.1 | - | 2 - 3 | 206 | 97 | |
| AK055916.1 | - | 2 - 3 | 910 | 97 | ||
| AK303507.1 | - | 4 - 5 | 511 | 97 | ||
| AY700780.1 | - | 6 - 7 | 766 | 97 | ||
| CR933661.1 | - | 17 - 18 | 4416 | 97 | ||
More Information
Set Membership: |
|

PRRC2B
Genomic Map