Assay Details
| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297767.1 | NP_001284696.1 | 1 - 2 | 245 | 88 | |
| NM_001297769.1 | NP_001284698.1 | 1 - 2 | 245 | 88 | ||
| NM_001297770.1 | NP_001284699.1 | 1 - 2 | 245 | 88 | ||
| NM_016067.3 | NP_057151.1 | 1 - 2 | 245 | 88 | ||
| GenBank mRNA | AF151892.1 | - | 1 - 2 | 158 | 88 | |
| AK002008.1 | - | 1 - 2 | 111 | 88 | ||
| AK026620.1 | - | 1 - 2 | 212 | 88 | ||
| BC005186.1 | - | 1 - 2 | 136 | 3839381 | 88 | |
| BE874586.1 | - | 1 - 2 | 87 | 88 | ||
| CA455563.1 | - | 1 | 122 | 88 | ||
| CN262115.1 | - | 1 - 2 | 159 | 88 | ||
| DA446717.1 | - | 1 - 2 | 246 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MRPS18C
Genomic Map