Assay Details
| Gene Symbol: REPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286612.1 | NP_001273541.1 | 11 - 12 | 2029 | 119 | |
| GenBank mRNA | BC021211.2 | - | 11 - 12 | 1649 | 4136619 | 119 |
More Information
Set Membership: |
|

REPS1
Genomic Map