Assay Details
| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098794.1 | NP_001092264.1 | 8 - 9 | 1796 | 63 | |
| GenBank mRNA | AB051546.1 | - | 8 - 9 | 1701 | 63 | |
| AK056468.1 | - | 8 - 9 | 1748 | 63 | ||
| AK291702.1 | - | 8 - 9 | 1721 | 63 | ||
| BC030825.1 | - | 8 - 9 | 1709 | 4811942 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175100.hg.1 | TC1100009962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |

FAM160A2
Genomic Map