Assay Details
| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003720.1 | 1 - 2 | 47 | 81 | ||
| GenBank mRNA | AY154892.1 | - | 1 - 2 | 47 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | histatin 1 | HTN1 | HIS1 | -8.5kb | Sense | Chr. 4 - 70050415 - 70058848 on NCBI Build GRCh38 |
| Downstream |
casein alpha s2-like B, pseudogene |
CSN1S2BP | CSN1S2B | 64.5kb | Sense | Chr. 4 - 70133604 - 70146704 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000378.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY154892.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04005131.hg.1 | TC04000378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005131.hg.1 | ENST00000451783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005131.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005130.hg.1 | TC04000378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005130.hg.1 | ENST00000451783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005130.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005129.hg.1 | TC04000378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005129.hg.1 | ENST00000451783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005129.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152988.hg.1 | TC0400007773.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400152985.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152985.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152985.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152985.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152985.hg.1 | TC0400007773.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | ENST00000512167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007773.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000451783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000503711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | ENST00000503813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | NR_003720.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152989.hg.1 | TC0400007773.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 218564 | - | - |

CSN1S2AP
Genomic Map