Assay Details
| Gene Symbol: LOC645485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135046.1 | 1 - 2 | 194 | 87 | ||
| GenBank mRNA | BC039526.1 | - | 1 - 2 | 194 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | caprin family member 2 | CAPRIN2 |
C1QDC1, EEG-1, EEG1, RNG140 |
Anti-sense | Chr. 12 - 30709552 - 30754994 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 941 | LINC00941 | 14.9kb | Sense | Chr. 12 - 30795681 - 30802711 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711933 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16544884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280227 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280221 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711947 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08280215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28711970 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 1717 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 260723 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 304805 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 427546 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 809978 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 858243 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 327108 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 646701 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 60108 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 234059 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 91325 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 968517 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 64397 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 282549 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 56737 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3740529 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52423 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P19503282 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P19503282 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3740529 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P19503282 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19503282 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 134599 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 48846 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503265 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39631567 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503261 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503284 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503254 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503274 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503282 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503269 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P19214870 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503249 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19503274 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19503282 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02597430 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19503261 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1560251_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007199.hg.1 | ENST00000500076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007199.hg.1 | ENST00000500076.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007199.hg.1 | ENST00000500076.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007199.hg.1 | ENST00000500076.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1560251_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560251_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | XM_932931_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560251_at9 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 713389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4070722 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_40325 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1692833 | BC039526.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1692833 | BC039526.1 | human_transcriptome |

LOC645485
Genomic Map