Assay Details
| Gene Symbol: ADARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018702.3 | NP_061172.1 | 5 - 6 | 1686 | 96 | |
| GenBank mRNA | AF034837.1 | - | 5 - 6 | 1694 | 96 | |
| BC137477.1 | - | 5 - 6 | 1507 | 9021477 | 96 | |
| BC140852.1 | - | 5 - 6 | 1507 | 9021720 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000969.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012176.hg.1 | ENST00000381312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012176.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012176.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012176.hg.1 | TC10000969.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165201.hg.1 | TC1000009541.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381310.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000381312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000469464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000474762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000477140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | ENST00000490172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009541.hg.1 | NM_018702.3 | Refseq |

ADARB2
Genomic Map