Assay Details
| Gene Symbol: TNRC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025277.1 | - | 2233 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1567 | LINC01567 | -147.6kb | Sense | Chr. 16 - 24661422 - 24671062 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 5 member 11 | SLC5A11 |
KST1, RKST1, SGLT6, SMIT2 |
26.9kb | Anti-sense | Chr. 16 - 24832784 - 24911711 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TNRC6A
Genomic Map