Assay Details
| Gene Symbol: TNRC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023148.1 | XP_016878637.1 | 5 - 6 | 3230 | 62 | |
| XM_017023151.1 | XP_016878640.1 | 5 - 6 | 3228 | 62 | ||
| GenBank mRNA | AK025696.1 | - | 1 - 2 | 170 | 62 | |
More Information
Set Membership: |
|

TNRC6A
Genomic Map