Assay Details
| Gene Symbol: SSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135823.1 | NP_001129295.1 | 2 - 3 | 277 | 65 | |
| NM_005086.4 | NP_005077.2 | 2 - 3 | 460 | 65 | ||
| XM_011520853.2 | XP_011519155.1 | 2 - 3 | 210 | 65 | ||
| XM_011520854.2 | XP_011519156.1 | 2 - 3 | 384 | 65 | ||
| XM_011520855.2 | XP_011519157.1 | 2 - 3 | 126 | 65 | ||
| GenBank mRNA | AF016028.2 | - | 2 - 3 | 360 | 65 | |
| AK092925.1 | - | 2 - 3 | 273 | 65 | ||
| AK304667.1 | - | 3 - 4 | 354 | 65 | ||
| AK314353.1 | - | 2 - 3 | 449 | 65 | ||
| AL136756.1 | - | 2 - 3 | 444 | 65 | ||
| BC062299.1 | - | 2 - 3 | 440 | 6376868 | 65 | |
| BP341300.1 | - | 2 - 3 | 460 | 65 | ||
| X89105.1 | - | 2 - 3 | 282 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003291.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12003112.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003112.hg.1 | ENST00000422622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003112.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | ENST00000422622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | ENST00000441067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003123.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003113.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003113.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003113.hg.1 | ENST00000422622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003113.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199577.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000537525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000540625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010187.hg.1 | ENST00000541940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199569.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199570.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199578.hg.1 | TC1200012610.hg.1 | Clariom D TC |

SSPN
Genomic Map