Assay Details
| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178562.4 | NP_848657.1 | 2 - 3 | 420 | 85 | |
| GenBank mRNA | AF276891.1 | - | 2 - 3 | 155 | 85 | |
| AY236849.1 | - | 2 - 3 | 244 | 85 | ||
| BC044244.1 | - | 2 - 3 | 384 | 5759132 | 85 | |
| HY012137.1 | - | 2 - 3 | 421 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07012511.hg.1 | TC07000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012511.hg.1 | ENST00000289407.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012511.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012512.hg.1 | TC07000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012512.hg.1 | ENST00000289407.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012512.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000498745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000498745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000498745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000498745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | ENST00000614309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009079.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | ENST00000496963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700164613.hg.1 | TC0700009079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700164612.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164612.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700164612.hg.1 | ENST00000486685.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700164612.hg.1 | NM_178562.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700164612.hg.1 | TC0700009079.hg.1 | Clariom D TC |

TSPAN33
Genomic Map