Assay Details
| Gene Symbol: SFT2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199344.2 | NP_955376.1 | 3 - 4 | 301 | 81 | |
| GenBank mRNA | AK290325.1 | - | 3 - 4 | 301 | 81 | |
| AL035297.1 | - | 3 - 4 | 310 | 81 | ||
| BC068098.1 | - | 3 - 4 | 297 | 6304345 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006289.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022795.hg.1 | TC01006289.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022795.hg.1 | ENST00000271375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022795.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022795.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177490.hg.1 | TC0100010525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000271375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000367829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000471981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | ENST00000630869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010525.hg.1 | NM_199344.2 | Refseq |

SFT2D2
Genomic Map