Assay Details
| Gene Symbol: SCN3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511610.2 | XP_011509912.1 | 3 - 4 | 567 | 149 | |
| XM_017004663.1 | XP_016860152.1 | 3 - 4 | 567 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011511610.2, XM_017004663.1 transcripts.
Set Membership: |
|

SCN3A
Genomic Map